missing translation for 'onlineSavingsMsg'
Learn More

EDTA dipotassium magnesium salt, Fluka™

Product Code. 15720229 Shop All Honeywell Fluka Products
Change view
Click to view available options
:
Quantity: 25 kg; Packaging: Fiberboard Box; Grade: Complexometric Titration
Product Code.
15720229 Quantity: 25 kg; Packaging: Fiberboard Box; Grade: Complexometric Titration
1 options
This item is not returnable. View return policy

Product Code. 15720229

Brand: Honeywell-Fluka 3335025KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For complexometry, ≥98%

  • Leading the industry for over 65 years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EDTA dipotassium magnesium salt
CAS 15708-48-2
Quantity 25 kg
Molecular Formula C10H12K2MgN2O8 · 2H2O
Synonym EDTA-K2Mg
Molecular Weight (g/mol) 426.74
Percent Purity 98%
Packaging Fiberboard Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.