missing translation for 'onlineSavingsMsg'
Learn More

EDTA dipotassium magnesium salt, For complexometry, ≥98%, Solstice

Product Code. 15613950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
6 x 100 g
50 kg
Unit Size:
100g
50kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613950 6 x 100 g Pack of 6
15693940 100 g 100g
15603950 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15613950 Supplier Honeywell-Fluka Supplier No. 333506X100G
  • ONLINE EXCLUSIVE PRICE
    430.00 €
    334.75 € / Pack of 6
    Save 95.25 €
    22% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for complexometry,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15708-48-2
Molecular Formula C10H12K2MgN2O8
Molecular Weight (g/mol) 390.714
MDL Number MFCD00054450
InChI Key MUEOBEUHFKBRJH-UHFFFAOYSA-J
Synonym dipotassium magnesium ethylenediaminetetraacetate, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-.kappa.o methyl glycinato-.kappa.n,.kappa.o 4--, dipotassium, oc-6-21, edta dipotassium magnesium salt monohydrate, ethylenediaminetetraacetic acid dipotassium magnesium salt, ethylenediamine-n,n,n',n'-tetraacetic acid dipotassium magnesium salt, edta, dipotassium magnesium salt, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, dipotassium, oc-6-21, magnesate 2-, n,n'-1,2-ethanediylbis n-carboxy-kappao methyl glycinato-kappan,kappao 4--, potassium 1:2 , oc-6-21, magnesium 2+ dipotassium ion ion 4-edta, dipotassium n,n'-ethylenebis n-carboxymethyl glycinato 4--n,n',o,o',on,on' magnesate 2-
PubChem CID 85061
IUPAC Name magnesium;dipotassium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Mg+2].[K+].[K+]

Specifications

CAS Max % 100.0000%
Physical Form Solid
Quantity 6 x 100 g
Linear Formula C10H12K2MgN2O8 · 2H2O
Formula Weight 426.74
Percent Purity >98%
Grade Not Available
Packaging Poly Bottle
Chemical Name or Material EDTA dipotassium magnesium salt, For complexometry, ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.