missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt 0.1 mol/L, BAKER ANALYZED™, J.T. Baker™

Product Code. 10006200 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10006200 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10006200 Supplier JT Baker Supplier No. 7126.1000
  • ONLINE EXCLUSIVE PRICE
    73.49 €
    67.26 € / 1 L
    Save 6.23 €
    8% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Laboratory Reagent

Specifications

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
Quantity 1 L
Synonym Edetic acid disodium salt , Disodium dihydrogen ethylenediaminetetraacetate , Ethylenediaminetetra-acetic acid disodium salt , Idranal III , Kestranal™ 2 S , Ethylenedinitrilotetra-acetic acid disodium salt
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular Weight (g/mol) 336.21
Concentration or Composition (by Analyte or Components) 0.1 mol/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.