Learn More
Edwards Fittings for BOC High Vacuum Pumps: Centering Ring
Click to view available options
Material:
Stainless Steel
Type:
NW40 Centering Ring
Description
- Aluminum centering rings manufactured from high-tech polymer
- Unique slotted design prevents gas bursts
- CX2 polymer can be used at temperatures up to 120°C and is unaffected by most common solvents
- Outgassing rate of the material is 6.6 × 10-8mbar l s-1cm-2
Specifications
Specifications
| For Use With | BOC high vacuum pump |
| Material | Stainless Steel |
| Type | NW40 Centering Ring |