missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EEF2K Recombinant Human Protein

EEF2K is a highly conserved serine/threoning kinase in the calmodulin signaling pathway.

Brand:  Invitrogen™ PR8535A

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15458673

  • 1826.00 € / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EEF2K activity is increased in cancers, including breast cancer.

Specifications

Specifications

NP_037434
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
100μg
RUO - research use only
EEF2K, eEF-2K
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
109.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Alpha Kinase Family
Recombinant
E. coli
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers