missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Efficient-Pro™ Feed 2

Product Code. 30284229
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30284229 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30284229 Supplier Gibco™ Supplier No. A5221401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gibco™ Efficient-Pro™ Feed 2 helps deliver superior product performance, improved product titer, better protein quality and lot-to-lot consistency for more control and predictability. Feed 2 is chemically defined for maximum feeding flexibility and pairs best with feeds for CHO-S cells.

  • A next generation CHO feed used in the production of monoclonal antibodies
  • Feed 2 offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
  • Formulated as a chemically-defined, animal-origin free cell culture feed used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
  • Free of growth factors and insulin to ensure batch-to-batch consistency and production efficiency
  • Designed to work optimally with Efficient-Pro™ Medium, Efficient-Pro™ Feed 2 has been formulated specifically for use in CHO-S and DG-44 cell lines
  • The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
  • System is easier to use with less solubility issues than other CHO media and feed offerings
  • Sustain cell viability for longer bioreactor runs & maximized titer
  • Demonstrated to improve titers in a DG-44 cell line up to 146% when paired with Efficient-Pro™ Medium
  • Improve downstream outputs such as protein glycosylation, charge-variants, & aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Increase flexibility with a range of format & packaging configurations
  • Decrease cell shock with simplified adaptation protocols
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in the dark at 2–8°C.

Cell Line CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Mammalian Cell Culture
Form Liquid
Green Features Sustainable packaging
Quantity 1000 mL
Shipping Condition Room temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.