missing translation for 'onlineSavingsMsg'
Learn More

Efinaconazole, TRC

Product Code. 30380593
Change view
Click to view available options
Quantity:
1 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380593 1 mg
30369857 100 mg
30373261 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380593 Supplier Toronto Research Chemicals Supplier No. TRCE4350701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164650-44-6
Molecular Formula C18 H22 F2 N4 O
Molecular Weight (g/mol) 348.39
Synonym 1-Piperidineethanol, α-(2,4-difluorophenyl)-β-methyl-4-methylene-α-(1H-1,2,4-triazol-1-ylmethyl)-, (αR,βR)-, 1-Piperidineethanol, α-(2,4-difluorophenyl)-β-methyl-4-methylene-α-(1H-1,2,4-triazol-1-ylmethyl)-, [R-(R*,R*)]-, (αR,βR)-α-(2,4-Difluorophenyl)-β-methyl-4-methylene-α-(1H-1,2,4-triazol-1-ylmethyl)-1-piperidineethanol, Efinaconazole, Jublia, KP 103, KP 103 (pharmaceutical)
IUPAC Name (2R,3R)-2-(2,4-difluorophenyl)-3-(4-methylidenepiperidin-1-yl)-1-(1,2,4-triazol-1-yl)butan-2-ol
SMILES C[C@@H](N1CCC(=C)CC1)[C@](O)(Cn2cncn2)c3ccc(F)cc3F

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H22F2N4O/c1-13-5-7-23(8-6-13)14(2)18(25,10-24-12-21-11-22-24)16-4-3-15(19)9-17(16)20/h3-4,9,11-12,14,25H,1,5-8,10H2,2H3/t14-,18-/m1/s1
Formula Weight 348.1762
Chemical Name or Material Efinaconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.