missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ EGR1/CEN5q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1475.00 € - 2430.00 €
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | early growth response 1 |
Gene Alias | AT225,G0S30,KROX-24,NGFI-A,TIS8,ZIF-268,ZNF225 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16033345
|
Abnova™
FG0213.100UL |
100 μL |
1475.00 €
100µL |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16043345
|
Abnova™
FG0213.200UL |
200 μL |
2430.00 €
200µL |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
AT225,G0S30,KROX-24,NGFI-A,TIS8,ZIF-268,ZNF225 | |
1958 | |
Liquid |
Tube | |
early growth response 1 | |
EGR1 | |
DNA/RNA |