missing translation for 'onlineSavingsMsg'
Learn More

Eicosane, 99%

Product Code. 10246640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
2g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10246640 2 g Glass bottle 2g
10144410 100 g Glass bottle 100g
10636734 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10246640 Supplier Thermo Scientific Acros Supplier No. 204370020
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 112-95-8 | C20H42 | 282.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-95-8
Molecular Formula C20H42
Molecular Weight (g/mol) 282.56
MDL Number MFCD00009344
InChI Key CBFCDTFDPHXCNY-UHFFFAOYSA-N
Synonym eicosane, n-eicosane, unii-3aya9kec48, ccris 663, nonadecane, methyl, 3aya9kec48, eicosane, analytical standard, octyldodecane, n-icosane
PubChem CID 8222
ChEBI CHEBI:43619
IUPAC Name icosane
SMILES CCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 36.0°C to 38.0°C
Color White
Density 0.7886g/mL
Boiling Point 220.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)18CH3
Quantity 2 g
Beilstein 01, 174
Specific Gravity 0.7886
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether
Formula Weight 282.54
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Eicosane
Show More Show Less
compliance-icons
Product Identifier
  • Eicosane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.