missing translation for 'onlineSavingsMsg'
Learn More

Eicosanedioic Acid, TRC

Product Code. 30370918
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370918 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370918 Supplier Toronto Research Chemicals Supplier No. TRCE4768601G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2424-92-2
Molecular Formula C20H38O4
Quantity 1 g
Synonym 1,18-Octadecanedicarboxylic Acid, 1,20-Eicosanedioic Acid, C 20, C 20 Fatty Acid, Eicosan-1,20-dioic Acid, Octadecamethylenedicarboxylic Acid, SL 20-90, SL 20-93, SL 20-99
InChI Formula InChI=1S/C20H38O4/c21-19(22)17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-18-20(23)24/h1-18H2,(H,21,22)(H,23,24)
SMILES OC(=O)CCCCCCCCCCCCCCCCCCC(=O)O
Molecular Weight (g/mol) 342.51
Formula Weight 342.277
Chemical Name or Material Eicosanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.