missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Eicosanoic acid, 98%

Product Code. 10714041 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10714041

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 506-30-9 | C20H40O2 | 312.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-30-9
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.54
MDL Number MFCD00002755
InChI Key VKOBVWXKNCXXDE-UHFFFAOYSA-N
Synonym arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate
PubChem CID 10467
ChEBI CHEBI:28822
IUPAC Name icosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 74.0°C to 77.0°C
Color White
Boiling Point 328.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)18CO2H
Quantity 1 g
Merck Index 15, 753
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in hot absolute alcohol, benzene,, chloroform and ether
Formula Weight 312.52
Percent Purity 99%
Physical Form Glistening Powder or Flakes
Chemical Name or Material Eicosanoic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.