missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eicosapentaenoic Acid-d5 (Technical), TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Eicosapentaenoic acid 19,19,20,20,20-d5 |
| CAS | 1197205-73-4 |
| Molecular Formula | C20 2H5 H25 O2 |
| Quantity | 1 mg |
| Synonym | 5,8,11,14,17-Eicosapentaenoic-19,19,20,20,20-d5 acid, (5Z,8Z,11Z,14Z,17Z)-, (5Z,8Z,11Z,14Z,17Z)-5,8,11,14,17-Eicosapentaenoic-19,19,20,20,20-d5 acid, Eicosapentaenoic acid-d5, Eicosapentaenoic acid (19,19,20,20,20-D5) |
| InChI Formula | InChI=1S/C20H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-19H2,1H3,(H,21,22)/b4-3-,7-6-,10-9-,13-12-,16-15-/i1D3,2D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)O |
| IUPAC Name | (5Z,8Z,11Z,14Z,17Z)-19,19,20,20,20-pentadeuterioicosa-5,8,11,14,17-pentaenoic acid |
| Molecular Weight (g/mol) | 307.48 |
| Formula Weight | 307.256 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction