missing translation for 'onlineSavingsMsg'
Learn More

EIDD 2801, TRC

Product Code. 30006375
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006375 50 mg 50mg
30004041 25 mg 25mg
30005900 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006375 Supplier Toronto Research Chemicals Supplier No. TRCE98579550MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2492423-29-5
Molecular Formula C13 H19 N3 O7
Molecular Weight (g/mol) 329.306
Synonym Uridine, 4-oxime, 5'-(2-methylpropanoate), (4Z)- (ACI), EIDD 1931-isopropyl ester, EIDD 2801, Lagevrio, MK-4482, Molnupiravir, β-D-N4 hydroxycytidine-5'-isopropyl ester, Uridine (4Z)-4-oxime 5'-(2-methylpropanoate)
IUPAC Name [(2R,3S,4R,5R)-3,4-dihydroxy-5-[(4Z)-4-hydroxyimino-2-oxopyrimidin-1-yl]oxolan-2-yl]methyl 2-methylpropanoate
SMILES CC(C)C(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C\C(=N\O)\NC2=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H19N3O7/c1-6(2)12(19)22-5-7-9(17)10(18)11(23-7)16-4-3-8(15-21)14-13(16)20/h3-4,6-7,9-11,17-18,21H,5H2,1-2H3,(H,14,15,20)/t7-,9-,10-,11-/m1/s1
Formula Weight 329.122
Chemical Name or Material Molnupiravir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.