missing translation for 'onlineSavingsMsg'
Learn More

EIF2AK2 (PKR) Recombinant Human Protein

EIF2AK2 (PKR) Recombinant Human Protein

Brand:  Invitrogen PV4821

436.84 EUR valid until 2023-03-27
Use promo code "20585" to get your promotional price.



Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10022913

  • 652.00 € / Each
Estimated Shipment: 12-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

EIF2AK2 (PKR) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses. EIF2AK2 plays a key role in antiviral defense mediated through its direct activation by double-stranded RNA produced by viral infection.
Specifications

Specifications

NP_002750.1
62.2 kDa
EIF2AK2 (PKR)
aa 252-551
GST Tag
Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Dry Ice
EIF2AK2
Baculovirus
Recombinant, Catalytic
Serine/Threonine Kinases
Human
9833
10 µg
PEK Kinase Family
Yes
Storage Buffer: 50 mM Tris, pH 7.5, 50% glycerol, 0.5 mM EDTA, 0.02% Triton X-100, 150 mM NaCl, 2 mM DTT
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Ser⁄Thr Kinases: Other
LanthaScreen™ Antibody, LanthaScreen™ Substrate
eukaryotic translation initiation factor 2-alpha kinase 2
Kinases & Inhibitors
Liquid
Kinase Assay
Signal Transduction
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers