missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EIF2AK2 (PKR), GST Tag Recombinant Protein

Product Code. p-7181058
Click to view available options
Quantity:
10 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10022913

Brand: Invitrogen™ PV4821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

EIF2AK2 (PKR) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses. EIF2AK2 plays a key role in antiviral defense mediated through its direct activation by double-stranded RNA produced by viral infection. PKR plays a key role in IFN induced-innate antiviral response, virus-induced apoptosis, cell growth & differentiation. PKR induces apoptosis by up-regulating Fas expression and mediates FADD/Caspase-8 death signaling pathway. Upon viral infection, it functions as a dual protein, sequentially activating both cell survival & cell death pathways using kinase independent and dependent strategies. PRKR regulates multiple pathways which include NF-kappaB activation, p53, p38, & PDGF signaling pathway. PKR has been implicated in tumor suppression & malignancy. To evade the antiviral effects of PKR, viruses have evolved multiple mechanisms, such as the inhibition of PKR by the non-structural protein (NS1) of the influenza virus. More recently, PKR has been implicated in several neurodegenerative diseases including Alzheimer, Huntington, and amyotrophic lateral sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19525
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 2mM DTT, 50% glycerol, 0.5mM EDTA, 150mM NaCl, 0.02% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 5610
Molecular Weight (g/mol) 62.2 kDa
Name Human EIF2AK2 (PKR), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2310047A08Rik; 4732414G15Rik; AI467567; AI747578; double stranded RNA activated protein kinase; dsRNA-activated kinase; EC 2.7.11.1; eIF-2 alpha; eIF2a Kinase; eIF-2A protein kinase 2; EIF2AK1; EIF2AK2; eukaryotic translation initiation factor 2 alpha kinase 2; eukaryotic translation initiation factor 2-alpha kinase 2; IFN- type I-induced and dsRNA-activated kinase; IFN-induced and double-stranded RNA-activated kinase; interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; interferon-inducible RNA-dependent protein kinase; MGC126524; OTTHUMP00000201320; P1/eIF-2A protein kinase; p68 kinase; PKR; PPP1R83; Prkr; protein kinase R; protein kinase RNA-activated; protein kinase, interferon inducible double stranded RNA dependent; Protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; serine/threonine-protein kinase TIK; T-cell viral integration site; Tik; Tyrosine-protein kinase EIF2AK2
Common Name PKR
Gene Symbol EIF2AK2
KinaseFamily PEK Kinase Family
Protein Length 252-551
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 252-551
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate
Target Entry EIF2AK2
Target Gene eukaryotic translation initiation factor 2-alpha kinase 2
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.