Learn More
Chemsplash Eka Coverall (Sterile and Irradiated)
Product Code. 30058394
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058394 2X-Large Pack of 25
30058261 Small Pack of 25
30058246 Medium Pack of 25
30058139 Large Pack of 25
30058196 X-Large Pack of 25
30058312 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058394 Supplier Chemsplash Supplier No. 2765.400.008
  • ONLINE EXCLUSIVE PRICE
    154.00 €
    138.39 € / Pack of 25
    Save 15.61 €
    10% Off
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Eka 55 Disposable Coverall 55 gsm has an elasticated back, hood, cuffs and ankles, and an adhesive cover flap for additional sealing.
Chemsplash Eka 55 Disposable Coverall
  • 55GSM Microporous Laminate Fabric
  • Elasticated 3-piece Hood, Back and Cuffs
  • Two Way Zip and Adhesive Zip Flap
  • Elasticated Ankles
  • Silicone, Latex Free and PFAS Free
  • Non Linting Fabric and Anti-Static
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
  • Available in sizes 4-6XL (Codes 2756, 2757 and 2758)
Description: The Chemsplash Eka 55 Disposable Coverall is made from 55GSM microporous laminated non-linting fabric. This Cat III Type 5B/6B coverall offers better value for applications where the suit is not used for prolonged periods but still requires high protection against liquid chemical splashes. It features an elasticated back, hood, cuffs, and ankles, with an adhesive cover flap for additional sealing. Chemsplash Eco fabric is Anti-Static to EN1149-5:2008 and non-linting, making it ideal for environments that must remain uncontaminated by garment fibers.
Specifications
Product Type Cleanroom Coveralls
Size 2X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)