Learn More
Chemsplash Eka Jacket A/S with Collar Zip and Adhesive Flap
GREENER_CHOICE
Product Code. p-200105905
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058475
Brand: Chemsplash 2606.400.004
Only null left
Add to basket
Add to basket
Disposable Zip Jacket are made of soft and microporous Chemsplash fabric. Strong and resistant to chemical splash and dust.
Chemsplash Disposable Zip Jacket
  • Chemsplash 55GSM Type PB 6B Fabric
  • Long Sleeves and Collar
  • Covered Zip
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting Fabric
  • PFAS Free
  • Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9
Description: Chemsplash Disposable Zip Jacket is made of soft and microporous Chemsplash fabric. It is strong and resistant to chemical splash and dust. Lint-free and anti-static in accordance with EN1149-5:2008. Chemsplash accessories are used to protect the wearer from the environment or the environment from the wearer. The range is designed to allow for maximum comfort and wearability. Conforms to ISO 14644-1:2015 Cleanroom Class 5 - 9.
Specifications
Product Type Short Lab Coat
Size Small
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
No. of Pockets None
Latex Free Yes
Autoclavable Autoclavable
Cleanroom Class Class 100-100,000 (ISO 5-8)