Learn More
Chemsplash Eka Trouser
GREENER_CHOICE
Product Code. 30058351
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30058351 Small Pack of 50
30058358 Medium Pack of 50
30058239 Large Pack of 50
30058170 X-Large Pack of 50
30058436 2X-Large Pack of 50
30058400 - Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058351 Supplier Chemsplash Supplier No. 2616.400.004
  • ONLINE EXCLUSIVE PRICE
    162.00 €
    160.00 € / Pack of 50
    Save 2.00 €
    1% Off
Estimated Shipment: 18-03-2027
Add to basket
Add to basket
Trousers with Elasticated Waist and Ankles are strong and resistant to chemical splash and dust.
Chemsplash Trousers with Elasticated Waist and Ankles
  • Chemsplash 55GSM Type PB 6B Fabric
  • Elasticated Waist and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting Fabric
  • PFAS Free
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: Chemsplash Trousers with Elasticated Waist and Ankles are made of soft and microporous Chemsplash fabric. They are strong and resistant to chemical splash and dust, lint-free, and feature anti-static properties under EN1149-5:2008. Chemsplash accessories are designed to protect the wearer from the environment or the environment from the wearer. This range has been crafted to offer maximum comfort and wearability. The trousers conform to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9 standards.
Specifications
Size Small
Gender Unisex
Product Type Trouser
Material Microporous Non-woven Polyethylene Laminate
Color White
Ankle Style Elastic
Seam Type Stitched
No. of Pockets None
Disposable Single-use
Latex Free Yes
Sterility Non-sterile