missing translation for 'onlineSavingsMsg'
Learn More

Manutan Ekwo three nest clean/dirty locker with plinth, blue, hasp lock, 1185mm wide

Product Code. 12847319
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12847319 Supplier Manutan Supplier No. A040315
Estimated Shipment: 05-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Strong single person locker with two vertical compartments and a hat shelf mounted on a plinth. Supplied in various colours, either mounted on four feet (for cleaning underneath) or on a plinth. Fitted with hat shelf, vertical central divider and a hanging rail in each compartment. Available with hasp lock (for use with a padlock - not supplied) or cylinder lock (supplied with two keys). Ventilation at top and bottom.

  • Three lockers supplied
  • Hasp lock
  • Colour of body: Grey
  • Colour of door: Blue
  • Dimensions (H x W x D): 1800mm x 1185mm x 500mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.