missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alanine Aminotransferase (ALT/GPT) Activity Kit

Product Code. 17902101
Click to view available options
Quantity:
96 tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 17902101

Brand: Invitrogen™ EEA001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Alanine Aminotransferase (ALT/GPT) Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of ALT activity in animal serum, plasma, tissue, cultured cells, and cell culture supernatant. ALT is an important pyridoxal phosphate dependent enzyme in the intermediate metabolism of glucose and proteins. Clinically, the activity of serum alanine aminotransferase is often used as a marker for alcoholic liver disease, liver cirrhosis, and acute viral hepatitis.

In this assay, ALT catalyzes the amino conversion reaction between alanine and α-ketoglutaric acid to produce pyruvic acid and glutamic acid. Phenylhydrazine is added to form phenylhydrazone with pyruvic acid. Phenylhydrazone is a reddish brown solution under alkaline conditions, and ALT activity can be calculated by measuring the OD values at 510 nm.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.75–72.3 IU/L
Assay Sensitivity 0.75 IU/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 50 min.
Interassay CV 9
Intraassay CV 5.3
Quantity 96 tests
Sample Volume 10 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Alanine Aminotransferase (ALT/GPT)
Total Assay Time 80 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.