missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aspartate Aminotransferase (AST/GOT) Activity Kit

Product Code. 17992091
Click to view available options
Quantity:
96 tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 17992091

Brand: Invitrogen™ EEA003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Aspartate Aminotransferase (AST/GOT) Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of aspartate aminotransferase activity in various sample types. AST/GOT is a key enzyme in nitrogen metabolism, which is widely found in plasma and body tissues, including liver, heart, skeletal muscle, kidney, brain, pancreas, lung, and erythrocytes. Changes in AST/GOT activity were found in acute pancreatitis, ischemic stroke, severe burns, periodontitis, acute renal disease, and motor neuron disease.

The assay is based on the ability of AST/GOT to enable alpha-ketoglutaric acid and aspartic acid to displace amino and keto groups to form glutamic acid and oxaloacetic acid. Oxaloacetic acid can decarboxylate itself to form pyroracemic acid during the reaction. Pyroracemic acid reacts with 2,4-dinitro phenyl hydrazine (DNPH) to form 2,4, dinitrophenylhydrazone, showing reddish-brown color in alkaline solution. The OD values can be measured to calculate the enzyme activity.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.1–72.3 IU/L
Assay Sensitivity 1.1 IU/L
Content And Storage Store at 2–8°C.

Detection Method Colorimetric
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Microplate Reader
Incubation Time 50 min.
Interassay CV 6.8
Intraassay CV 5.3
Quantity 96 tests
Sample Volume 10 μL
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Aspartate Aminotransferase (AST/GOT)
Total Assay Time 80 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.