missing translation for 'onlineSavingsMsg'
Learn More

PL BioScience ELAREM™ Perform-FD, Human Platelet Lysate, Fibrinogen-depleted, No Heparin Required

Product Code. 17907731
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Purity or Quality Grade:
GMP Grade
Research Grade
Unit Size:
100mL
500mL
50mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Purity or Quality Grade Unit Size
17907731 50 mL Research Grade 50mL
17967741 50 mL GMP Grade 50mL
17997731 100 mL Research Grade 100mL
17997721 100 mL GMP Grade 100mL
17937731 500 mL Research Grade 500mL
17927731 500 mL GMP Grade 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17907731 Supplier PL BioScience Supplier No. PE20612
  • ONLINE EXCLUSIVE PRICE
    220.39 €
    195.00 € / 50mL
    Save 25.39 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ELAREM™ Perform-FD is a fibrinogen-depleted human Platelet Lysate of US origin. As a cell culture supplement, it supports in vitro expansion of various primary cells and cell lines. Due to the fibrinogen-depletion process, ELAREM Perform-FD does not require anticoagulant addition.

  • Xeno-Free: The xeno-free cell culture supplement is obtained from transfusion-approved platelets obtained at licensed blood centres.
  • No Heparin Addition Required: Since fibrinogens are depleted during manufacture, coagulation of the complete medium is inhibited– application workflows are simplified. This eliminates the need to add an anticoagulant with ELAREM™ Perform-FD. Traces of a xeno-free anticoagulant may be present in the Human Platelet Lysate.
  • US Origin: ELAREM™ Perform-FD is a fibrinogen-depleted Human Platelet Lysate of US origin supporting in vitro expansion of various primary cells and cell lines.
  • Improved Cell Performance: Human Platelet Lysate contains abundant growth factors and cytokines derived from human platelets. Those nutrients have been shown to stimulate cellular proliferation and maintain phenotype and differentiation potential of various cells.
  • Reproducible Results: Each batch of ELAREM™ Perform is produced from large pools of transfusion-approved platelet units to ensure batch-to-batch consistency and enable reproducible conditions. Grade & GMP GradeB
  • From Research to Clinical Development: ELAREM™ Perform meets the requirements of academic and industrial cell expansion and assures an easy transition of research results into clinical applications.
  • The product contains coagulation factors. To inhibit coagulation of the complete cell culture medium, it is recommended to add an anticoagulant, e.g. heparin, to the cell culture medium.
  • Intended Use: ELAREM™ Perform is for in vitro and manufacturing use only. The product is not intended for direct therapeutic use. For safety precautions, please see appropriate Safety Data Sheet (SDS).

Specifications

Color Yellow
Culture Type Adherent Cell Culture, Suspension Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Sterility Sterile
Sterilization Method Sterile-filtered
Formulation Liquid, Human Platelet Lysate, Fibrinogen-depleted
Purity or Quality Grade Research Grade
Quantity 50 mL
Shipping Condition Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.