missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Electric Circular Boiling Bath
Shop All Nickel-Electro Products
Click to view available options
Capacity (Metric):
3.24 L
Unit Size:
Each
Description
- The steam produced provides an effective source of heat for samples:
- that may be otherwise thermally damaged by a naked flame
- used for applications to distill.concentrate liquids, melt agar and other solids
- metallurgical analysis, protein analysis and bacteriological examinations
- Features an integral constant level device to maintain constant water level during boiling and supplied with a set of rings to vary the diameter of aperture to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks.
- Features a built-in energy regulator, which controls the amount of boiling in a bath from a gentle simmer to a vigorous boil.
- Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
- The bath provides constant boiling water for gentle heating of samples in steam. Always site the Boiling Bath on a heat proof surface.
Safety Features:
- Always connect the constant level with laboratory tubing to a water supply, adjust the flow to gently top up the water level, whilst maintaining boiling.
- The Constant water level has a visible water spout for checking water is topping up the Boiling bath in use and avoids the chances of boiling dry.
- Manual reset cut out.
Specifications
Specifications
| Chamber Material | Stainless Steel |
| Controller Type | Energy regulator |
| Capacity (Metric) | 3.24 L |
| Temperature Range | Simmer to boiling |
| Material | Stainless Steel |
| Model | 1026 ER |
| Shape | Circular |
| Warranty | 3 Years |
| Depth (Metric) | 127 mm |
| Diameter (Metric) | 203 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction