missing translation for 'onlineSavingsMsg'
Learn More

BICASA Electric MECC Panel with 4 Multiple Sockets, Automatic Breaker

Product Code. 17457383 Shop All BICASA Products
Change view
Click to view available options
Electric MECC Panel with 4 Multiple Sockets, Automatic Breaker
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17457383 Electric MECC Panel with 4 Multiple Sockets, Automatic Breaker
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17457383 Supplier BICASA Supplier No. MBEEITA
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Electric MECC Panel with 4 Multiple Sockets (IP65) 250V/16A/2P+E and 1 automatic breaker 2P/16A.

  • Installed directly on the MECC Service Spine
  • PP resin moulded frontal panel - V0 flame retardant (UL 94)
  • Electric panels WITH SOCKETS
    • Sealed flame retardant ABS rear box with cable gland
    • Equipped with wiring and flexible power supply cable
    • FG7 (O) M1 self-extinguishing and halogen-free
  • 4 multiple sockets (IP65) 250V/16A/2P+E
  • 1 automatic breaker 2P/16A

Specifications

Count (Outlets) 4
For Use With (Equipment) MECC Series Service Spine
Voltage 230 V
Amperage 16 A
Color Gray
Includes 1 Automatic Breaker (2 P/16 A)
Outlet Type Multiple Sockets (IP65)
Frequency 50
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.