missing translation for 'onlineSavingsMsg'
Learn More

Erlab Electrical Base Wheels, 198 cm

Product Code. 31794137 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
Solis 1500
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31794137 Solis 1500 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31794137 Supplier Erlab Supplier No. NORT000431
  • ONLINE EXCLUSIVE PRICE
    6225.00 €
    5809.00 € / Pack of 1
    Save 416.00 €
    6% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The electric high-rise base allows the working position to be adjusted to the user’s specific needs, whilst providing a stable and ergonomic solution for high-level set-ups.

  • Benches, Rolling Carts and Bases for Erlab Enclosures: Erlab benches and rolling carts are designed to supPort ductless enclosures and filtering cabinets at the correct working height, in complete safety. Each model is dimensioned for a specific enclosure reference to guarantee stability and a perfect match with the equipment footprint.
  • Stability and Ergonomics:
    • Robust steel structure sized for the load of the corresponding enclosure
    • Chemically resistant, easy-to-clean finish
    • Fixed, mobile, levelling-feet, mechanically adjustable and electrically adjustable versions available
    • Height-adjustable models allow the working position to be adapted to each operator, seated or standing
    • Lockable castors on rolling versions for safe repositioning of the workstation
  • Flexible Laboratory Layout: Rolling carts make it possible to move a complete filtration workstation from one room to another as projects evolve, while fixed benches provide a permanent, space-efficient installation. Optional accessories such as retractable shelves and footrests further improve daily comfort and storage capacity.
  • Key Benefits:
    • Dedicated supPort designed for a specific Erlab enclosure.
    • Stable, robust structure with chemically resistant finish.
    • Fixed, mobile and height-adjustable versions available.
    • Improved operator ergonomics and workstation flexibility.
    • Simple assembly and easy maintenance.

Specifications

Product Type Electrical Base Wheels
For Use With (Equipment) Solis 1500
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.