Learn More
Mettler Toledo™ Electrode Arm, EasyPlace Complete
Product Code. 17531016
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17531016 EasyPlace Each
1 options
Product Code. 17531016 Supplier Mettler Toledo™ Supplier No. 30671569
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The EasyPlace electrode arm supports efficient measurement and calibration. With perfect vertical movement, spillage risks are minimized. Includes parking beaker, parking beaker insert and sachet holder.
  • Precise Control
  • Perfectly vertical sensor movement makes it easy to insert the sensor into samples
  • The right place
  • The EasyPlace contains dedicated positions for calibration, short- and long term sensor storage and measurement, simplifying everyday workflows
  • Fits right in
  • The EasyPlace electrode arm fits together with the EasyMix magnetic stirrer and standard sachets, simplifying operation and saving space
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode Arm, EasyPlace Complete
For Use With (Equipment) EasyPlace