missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Electrode Combined Surfactant

Product Code. 11759718 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Temperature (Metric) Operating:
0°C to 50°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature (Metric) Operating Unit Size
11759718 0°C to 50°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11759718
  • ONLINE EXCLUSIVE PRICE
    636.45 €
    344.57 € / Pack of 1
    Save 291.88 €
    45% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Combined ion-sensitive electrode with solid membraneinterchangeable flat surface, is ideal for measuring temperatures between 0 to 50 ° C.

  • Combined ion-sensitive electrode with solid membrane Interchangeable flat surface
  • The ion to be detected passes through the membrane generating a difference of potential proportional to ionic concentration
  • Analysis time generally less than one minute
  • Sample preparation steps such as filtering or distillation rarely necessary
  • Calibration should be carried out as often as possible (less once a day) at the same temperature as the sample
  • Measurement temperature 0 to 50 ° C

Specifications

Description Combined ion-sensitive electrode with solid membrane
Membrane Solid Membrane
Includes Membrane, electrolytic solution and gasket.
Temperature (Metric) Operating 0°C to 50°C
Type Combined ISE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.