missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA-Light™ Immunoassay System with CDP-Star™ and Emerald-II™ Substrate/Enhancer Solution

Product Code. 12644285
Click to view available options
Quantity:
1,000 assays
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12644285

Brand: Invitrogen™ T1026

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ultra-sensitive detection of antigens or antibodies in multiple immunoassay formats employing an alkaline phosphatase enzyme label.

  • Heterogeneous assay format increases sensitivity and dynamic range and reduces interference by test compounds in the detection portion of the assay (as compared with homogeneous assay formats)
  • Chemiluminescent immunoassay detection allows greater sensitivity and improved dynamic range as compared to radioimmunoassays and immunoassays that employ colorimetric or fluorescent detection
  • High sensitivity and wide dynamic range permit sensitive detection with fewer assays, because sample dilution to fit the dynamic range of the assay is unnecessary
  • A variety of ready-to-use substrate formulations lets the user precisely optimize signal intensity to the exact requirements of an assay
  • The kinetics of the glow-assay provides a window during which measurements may be performed, facilitating HTS applications where assay automation is used
  • Chemiluminescent immunoassay detection provides versatile applications such as sandwich immunoassays, competitive immunoassays, and DNA probe capture assays
  • Non-radioactive immunoassays eliminate concerns over use of radioisotopes

Extend the Limits of Detection
The ELISA-Light system offers an excellent combination of high sensitivity and throughput for assays measured in a luminometer. The ELISA-Light system significantly extends the limits of detection of most immunoassays previously performed with colorimetric and isotopic methods.

Multiple Substrate/Enhancer Formulations
The kit contains reagents optimized for chemiluminescent ELISAs employing alkaline phosphatase labels and is available with a choice of ready-to-use chemiluminescent substrate formulations containing CSPD or CDP-Star™ substrate with Sapphire-II or Emerald-II™ luminescence enhancers. Multiple substrate/enhancer formulations provide options that let the user optimize specific assays or luminometers. The specific formulation that will provide the highest sensitivity for a specific assay can vary. Therefore, an ELISA sampler kit containing all four substrate/enhancer formulations is available to facilitate assay development.

Specifications

  • Detection Method: Chemiluminescent
  • For Use With (Equipment): Microplate Reader
  • Label or Dye: AP Alkaline Phosphatase
TRUSTED_SUSTAINABILITY

Specifications

Product Type Immunoassay System
Quantity 1,000 assays
Detection Method Chemiluminescence
Conjugate AP
For Use With (Equipment) Microplate Reader
Product Line ELISA-Light
Substrate Type Alkaline Phosphatase
Content And Storage This kit contains enough reagent for 1000 assays. Store at 4°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.