missing translation for 'onlineSavingsMsg'
Learn More

ELISA Systems™ Egg Residue Detection Kit for Food Allergen Testing

Product Code. 13205699
Change view
Click to view available options
Quantity:
48 Tests
Unit Size:
48 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13205699 48 Tests 48 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13205699 Supplier Thermo Scientific Oxoid Supplier No. ESERD48
  • ONLINE EXCLUSIVE PRICE
    914.55 €
    854.96 € / 48 tests
    Save 59.59 €
    6% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect and quantify egg material in uncooked food and wine.

Simplify allergen testing with ELISA Systems™ Egg Residue Detection Kit which helps ensure compliance with food product labeling requirements. The enzyme-linked immunosorbent assay is designed to deliver simple, sensitive, and fast detection and quantification of casein as an indicator of the presence of milk or milk products. This complete, ready-to-use kit is developed and optimized for rapid and reliable test results that significantly reduce the time required to screen appropriate food products for the presence of egg residue.

Hen’s egg is one of the more frequent causes of food hypersensitivity in infants and young children.1, 2

Use ELISA SystemsEgg Residue Detection Kit for the rapid detection and quantification of egg residue in food and environmental samples. The kit is primarily suitable for the detection of residues of uncooked egg and is also suitable for wine testing.

  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting


Two of the main allergens in egg white are Ovalbumin (54%) and Ovomucoid (11%)3. Ovalbumin is the major allergen present in the egg white. Ovomucoid is allergenic in minute quantities and is not coaguable by heat4.

Ovalbumin (because of its allergenic properties) and Ovomucoid (because of its allergenic and heat stable properties) were chosen as the Egg protein indicators for the ELISA Systems Egg Residue Detection Kit.

All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.
TRUSTED_SUSTAINABILITY

Specifications

Description ELISA Systems Egg Residue Detection Kit
Detectable Analytes Ovalbumin and Ovomucoid Egg Protein
Immunoassay Kit Format ELISA (Standard)
Product Type Egg Residue Detection Kit
Kit Contents 20X Extraction solution concentrate, 20X wash buffer solution concentrate, Enzyme conjugate, Negative Standard, Positive Standards, Stop solution, Substrate, Test strips
Quantity 48 Tests
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.