missing translation for 'onlineSavingsMsg'
Learn More

ELISA Systems™ Hazelnut Residue Detection kit for Food Allergen Testing

Product Code. 13215699
Change view
Click to view available options
Quantity:
48 Tests
Unit Size:
48 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13215699 48 Tests 48 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13215699 Supplier Thermo Scientific Oxoid Supplier No. ESHRD48
  • ONLINE EXCLUSIVE PRICE
    914.55 €
    854.96 € / 48 tests
    Save 59.59 €
    6% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly detect and quantify hazelnut material in food and environmental samples.

Simplify allergen testing with ELISA Systems™ Hazelnut Residue Detection Kit which helps ensure compliance with food product labeling requirements. The enzyme-linked immunosorbent assay is designed to deliver simple, sensitive, and fast detection and quantification of hazelnut material in food and environmental samples. This complete, ready-to-use kit is developed and optimized to specifically detect heat stable hazelnut protein in order to avoid false negative results in highly heat-treated samples.

The Hazelnut (Corylus avellana) belongs to a group of foods, commonly referred to as “tree nuts”. This group of foods is a significant source of allergens and hazelnuts are a common cause of food allergy. Allergic reactions to hazelnuts range from oral allergy syndrome to severe anaphylactic reactions.1

Use ELISA Systems Hazelnut Detection Kit for the rapid detection and quantification of hazelnut material in food and environmental samples.

  • Fast: Sample preparation to result in under 60 minutes for rapid test turnaround and actions
  • Simple: Ready-to-use liquid reagents and concentrates, plus common handling procedure for all kits, make ELISA Systems Assays easy to implement and run
  • Robust: Ready-to-use liquid controls and allergen-free non-control reagents mean reduced risk of cross contamination that could lead to false-positive results
  • Flexible: Use for qualitative or quantitative testing—a single kit for more applications
  • Sensitive: Detect proteins or protein residues, including heat stable proteins, at low levels for improved assurance of product safety
  • Supported: Our experienced technical team is ready to assist you and help mitigate delays in test reporting


A sensitive and reliable test for the detection of hazelnut in foods must be able to detect a heat stable protein component; otherwise, false negative results could occur with samples that have been highly heat-treated.

A specific heat-stable hazelnut protein was identified and is used as the hazelnut protein indicator for the ELISA Systems Hazelnut Residue Detection kit. Results are expressed as ppm (mg/kg) hazelnut protein.

All results should be interpreted as part of a HACCP plan for food allergens. Any sample returning a positive result should be regarded as a presumptive result, and confirmation or further testing should be performed.

Not all products are available for sale in all territories. Please inquire.

ELISA Systems™ Food Allergen Test Kits are manufactured by ELISA Systems Pty Ltd.

ELISA Systems is a trademark of ELISA Systems Pty Ltd.
TRUSTED_SUSTAINABILITY

Specifications

Description Elisa Systems Hazelnut Residue Detection Kit
Detectable Analytes Heat Stable hazelnut protein
Immunoassay Kit Format ELISA (Standard)
Product Type Hazelnut Residue Detection Kit
Kit Contents 20X Extraction solution concentrate, 20X Extraction Solution concentrate for samples containing polyphenols, 20X wash buffer solution concentrate, Enzyme conjugate, Negative Standard, Positive Standards, Stop solution, Substrate, Test strips
Quantity 48 Tests
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.