missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Ellipticine hydrochloride

Product Code. 16446916
Encompass_Preferred
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16446916 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16446916 Supplier MedChemExpress Supplier No. HY15753A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ellipticine (NSC 71795) hydrochloride is a potent antineoplastic agent; inhibits DNA topoisomerase II activities.

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 282.77 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 5081-48-1
Product Type Natural Products
Purity 99.34%
Solubility DMSO : 5.8 mg/mL (20.51 mM∣Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic∣warming∣heat to 60°C) (insoluble)
Synonym NSC 71795 hydrochloride
Target Topoisomerase
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.