missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Elplasia™ 12K Flask

Product Code. 17811965 Shop All Corning Products
Change view
Click to view available options
Culture Area:
80 cm2
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area Unit Size
17811965 80 cm2 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17811965 Supplier Corning™ Supplier No. 4537
  • ONLINE EXCLUSIVE PRICE
    535.00 €
    496.61 € / Pack of 5
    Save 38.39 €
    7% Off
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Elplasia™ 12K Flask enables researchers to generate a high density of animal-free spheroids in a scaffold-free model.

  • Contains 152 microcavities per cm2 in a vessel footprint similar to that of a T-75 flask
  • Corning™ Ultra-Low Attachment (ULA) surface works with gravity and a rounded microcavity geometry to form approximately 12,000 spheroids of similar shape and size
  • ULA surface (a proprietary, animal-free, covalently bonded hydrogel), promotes the formation and easy harvesting of spheroids
  • Polystryene film is gas permeable with low autofluorescence
  • Round well-bottom geometry
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Orange
Material Polystyrene
For Use With (Application) Spheroid Formation
Sterility Sterile
Bottom Shape Rectangular
Closure Type Vented Cap
Culture Area 80 cm2
Volume (Metric) Working 25 to 50 mL
Quantity 5 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.