missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Elplasia™ Microcavity Plates
BIOPHARMA

Product Code. 16425289 Shop All Corning Products
Change view
Click to view available options
Well Count:
6
24
Surface Treatment:
Low Cell Bind
Tissue Culture
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Count Surface Treatment Unit Size
16425289 6 Low Cell Bind Pack of 5
16445299 24 Low Cell Bind Pack of 5
16465299 6 Tissue Culture Pack of 5
16475299 24 Tissue Culture Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16425289 Supplier Corning™ Supplier No. 4440
  • ONLINE EXCLUSIVE PRICE
    655.00 €
    608.67 € / Pack of 5
    Save 46.33 €
    7% Off
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The only plates with a simple 'plug and play' protocol for scaffold-free, self-assembly spheroid formation at large volumes.

  • Novel well geometry and surface coatings, including Corning Ultra Low Attachment (ULA) featured on the round bottom plates
  • Corning ULA surface is hydrophilic, biologically inert and non-degradable, which promotes highly reproducible spheroid formation and easy harvesting
  • Corning Elplasia round bottom plates optimal for bulk spheroid formation, collection and expansion\
  • Round bottom plates feature Corning Ultra-Low Attachment surface and are available in 6-, 24- and 96-well formats
  • Corning Elplasia square bottom plates feature a surface with optical qualities suited for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters
  • The Elplasia square plate is well suited for screening programs as it generates a high volume of spheroids per well, allowing for higher signals per well
  • Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats
  • Spheroids may be formed and cultured for 21 or more days
  • Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
  • The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
  • Black opaque sidewalls to reduce well-to-well 'cross-talk'
  • Well suited for fluorescent/luminescent assays
  • Available in multiple formats, two well geometries and surface coating options
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Round
Capacity (Metric) 16.8 mL
Color Black, Clear
Material Polystyrene
Shape Rectangular
Sterility Sterile
Surface Treatment Low Cell Bind
Type Microcavity Plate
Well Count 6
Includes Lid
Quantity 5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.