missing translation for 'onlineSavingsMsg'
Learn More

Eluxadoline, TRC

Product Code. 30373192
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373192 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373192 Supplier Toronto Research Chemicals Supplier No. TRCE50819510MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eluxadoline
CAS 864821-90-9
Molecular Formula C32 H35 N5 O5
Quantity 10 mg
Synonym Benzoic acid, 5-[[[(2S)-2-amino-3-[4-(aminocarbonyl)-2,6-dimethylphenyl]-1-oxopropyl][(1S)-1-(5-phenyl-1H-imidazol-2-yl)ethyl]amino]methyl]-2-methoxy-, Benzoic acid, 5-[[[(2S)-2-amino-3-[4-(aminocarbonyl)-2,6-dimethylphenyl]-1-oxopropyl][(1S)-1-(4-phenyl-1H-imidazol-2-yl)ethyl]amino]methyl]-2-methoxy- (9CI), 5-[[[(2S)-2-Amino-3-[4-(aminocarbonyl)-2,6-dimethylphenyl]-1-oxopropyl][(1S)-1-(5-phenyl-1H-imidazol-2-yl)ethyl]amino]methyl]-2-methoxybenzoic acid, Eluxadoline, JNJ 27018966, Viberzi
InChI Formula InChI=1S/C32H35N5O5/c1-18-12-23(29(34)38)13-19(2)24(18)15-26(33)31(39)37(17-21-10-11-28(42-4)25(14-21)32(40)41)20(3)30-35-16-27(36-30)22-8-6-5-7-9-22/h5-14,16,20,26H,15,17,33H2,1-4H3,(H2,34,38)(H,35,36)(H,40,41)/t20-,26-/m0/s1
SMILES COc1ccc(CN([C@@H](C)c2nc(c[nH]2)c3ccccc3)C(=O)[C@@H](N)Cc4c(C)cc(cc4C)C(=O)N)cc1C(=O)O
IUPAC Name 5-[[[(2S)-2-amino-3-(4-carbamoyl-2,6-dimethylphenyl)propanoyl]-[(1S)-1-(4-phenyl-1H-imidazol-2-yl)ethyl]amino]methyl]-2-methoxybenzoic acid
Molecular Weight (g/mol) 569.65
Formula Weight 569.2638
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.