missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Thermo Scientific™ E. coli DNA Ligase is an NAD-dependent DNA ligase. It catalizes formation of phosphodiester bonds between 5`-phospate and 3`-hydroxyl termini in double-stranded DNA. The enzyme is not active on blunt-ended DNA.
Features
- Activity in wide temperature range (4°C to 37°C)
- High specificity to cohesive DNA ends
Applications
- Nick ligation in double-stranded DNA
- cDNA synthesis
- Cloning
Specifications
Specifications
| Concentration | 8 U/μL |
| For Use With (Application) | Nick ligation in double-stranded DNA, cDNA synthesis, Cloning, DNA ligation |
| Content And Storage | 8 U/μL E.Coli DNA Ligase, 50 μL Store at -20°C. |
| Enzyme | DNA Ligase |
| Sensitive to Heat Inactivation | Yes |
| Quantity | 400 U |
| Product Type | NAD-dependent DNA Ligase |
| Research Category | Traditional Cloning |
| Inactivation Temperature | 65°C |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?