missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Select MALAT1 Positive Control siRNA

Product Code. 15667718
Change view
Click to view available options
Quantity:
5 nmol
Unit Size:
5nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15667718 5 nmol 5nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15667718 Supplier Invitrogen™ Supplier No. 4455877
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silencer™ Select MALAT1 Positive Control siRNA is extensively validated and an ideal control for many aspects of an siRNA experiment. 5 nmol is provided.

Silencer™ Select MALAT1 Positive Control siRNA is extensively validated and an ideal control for many aspects of an siRNA experiment. 5 nmol is provided.

  • Premium-quality siRNA, highly purified and ready to use
  • Functionally tested in several common cell lines
  • Include Silencer™ Select modifications for enhanced specificity
  • For use in human, mouse, and rat cells

What are Silencer™ Select siRNAs

Silencer™ Select siRNAs incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemical modifications, to yield siRNAs with unrivalled efficacy, potency, and specificity. The result is fewer failed experiments due to poor silencing, and cleaner, more consistent phenotypic data.

Silencer™ Select MALAT1 Positive Control siRNA

The Silencer™ MALAT1 positive control is extensively validated and an ideal 'test' siRNA for those just beginning siRNA experiments, because it is validated to work in multiple cell lines. Because it targets MALAT1 mRNA, many assays are available to measure its effects, including real-time RT-PCR.

Quality Control:

Silencer™ Select siRNAs are manufactured in a state-of-the-art facility under the highest quality standards. Rigorous quality control procedures include MALDI-TOF mass spectrometry analysis and analytical HPLC to monitor purity. Each siRNA is also assessed by gel electrophoresis to confirm that the strands anneal properly.

Accessory Product: siPORT™ NeoFX™ Transfection Agent (SKU #AM4510 and #AM4511)

siPORT™ NeoFX™ Transfection Agent is a versatile, lipid-based transfection agent for the efficient transfection of siRNA into cultured cells. The protocol can be adapted to a wide range of cells and experimental designs, including high-throughput applications.

Silencer™ Select siRNA controls have specific chemical modifications; they are designed to be used with Silencer™ Select siRNAs. If you are using other siRNA products, such as BLOCK-iT™ siRNAs, Ambion™ Silencer™ siRNAs, or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.

Lipofectamine™ RNAiMAX™ Transfection Reagent (SKU 13778075)

Lipofectamine™ RNAiMAX™ Transfection Reagent is a leading transfection reagent designed specifically for RNA interference applications. The protocol can be adapted to a wide range of cells and experimental designs, including high-throughput applications. Silencer Select siRNA controls have specific chemical modifications that are designed to be used with Silencer Select siRNAs. If you are using other siRNA products, such as Silencer siRNA or Stealth RNAi siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The siRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C.
Control Type Positive Control
RNAi Type siRNA
For Use With (Application) Transfection, RNAi
Label or Dye Unconjugated
Product Line Silencer Select, Ambion
Product Type siRNA
Purity HPLC
Quantity 5 nmol
Shipping Condition Dry Ice
Target MALAT1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.