missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Embryonic Stem Cell FBS, qualified, US origin
GREENER_CHOICE

Product Code. 15946942
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Format:
Bottle
One Shot
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 15946942

Brand: Gibco™ 16141002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fetal Bovine Serum, ES cell qualified, is specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells.

Fetal Bovine Serum, ES cell qualified, is specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells. It is Gibco FBS quality and performance you trust offered in a single-use FBS aliquot, which provides greater convenience, saves time, and is easy to use.

  • Sera Category: Specialty

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Fetal
Country of Origin United States
Form Liquid
Format One Shot
Product Type Fetal Bovine Serum
Quantity 50 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Tested For Bovine Respiratory Syncytial Virus FA, Cytopathic Agents, Hemadsorbing Agents, Reovirus FA, Rabies FA, Hemoglobin (ng/dL), Blue Tongue Virus, Bovine Adenovirus FA, Bovine Parvovirus FA, Osmolality, Sp2 Performance Assay, Electrophoretic Pattern, Total Protein, Endotoxin (EU/mL), Mycoplasma, Sterility Testing (Bacteria and Fungi)
Donor Herd No
Endotoxin Concentration <50 EU/mL
Hemoglobin Concentration ≤25 mg/dL
Purity or Quality Grade ES Cell-Qualified
Product Line One Shot
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.