missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™ Block Noise Canceling Reagents

Product Code. 11320250 Shop All Merck Products
Change view
Click to view available options
For Use With (Application):
Fluorescence Assay
Mass Spectrometry
Product Type:
Block-CH Reagent
Block-FL Reagent
Block-PO Reagent
Unit Size:
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Product Type unitSize
11320250 Fluorescence Assay Block-FL Reagent 500mL
13141294 Mass Spectrometry Block-CH Reagent 500mL
11340250 Mass Spectrometry Block-PO Reagent 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11320250 Supplier Merck Supplier No. WBAVDFL01
  • ONLINE EXCLUSIVE PRICE
    205.00 €
    188.67 € / 500mL
    Save 16.33 €
    7% Off
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Minimize background in Western blots, increasing sensitivity for superior detection of even low-abundance proteins

 

  • Ready-to-use
  • Protein-free, reducing nonspecific binding, especially when using phosphoantibodies or avidin-biotin systems
  • Specially engineered for the EMD Millipore SNAP i.d.™ detection system, but may be used in traditional Western blotting protocols
  • Suitable for the dilution of primary and secondary antibodies
  • Sufficient for 19 miniblots (each 8 × 7cm blot requires at least 26mL for SNAP i.d.)
  • Compatible with streptavidin antibodies
  • Compatible with PVDF (Immobilon™ P, Immobilon FL) and nitrocellulose

 

 

  • Block-CH reagent is compatible with Immobilon HRP substrate
  • Choose from three Block reagents
  • Block-CH reagent for HRP chemiluminescence detection
  • Block-FL reagent for fluorescence detection
  • Block-PO reagent for phosphoprotein detection via chemiluminescence or fluorescence

 

TRUSTED_SUSTAINABILITY

Specifications

Product Type Block-FL Reagent
For Use With (Application) Fluorescence Assay
Quantity 500 mL
Sufficient For Nineteen 8 x 7cm miniblots
Content And Storage 20°C to 25°C
compliance-icons compliance-icons
Product Identifier
  • Immobilon Block - FL (Fluorescent Blocker)
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-methyl-2H-isothiazol-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.