missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipore Diffusion Chamber Rings
Plexiglas rings with 14mm dia. for use with Diffusion chamber
Brand: Merck Millipore PR0001400
Code : 72
Additional Details : Weight : 1.00000kg
Description
- Available with or without hole
Specifications
Plexiglas™ Rings | |
Diffusion Chamber Kit | |
0.55 in. |
Plexiglas™ | |
14mm | |
No hole |