Learn More
Merck Millipore EZ-Fit™ Manifold 6-Place for Rubber Stopper Tulip
For use in general microbiological analysis
Brand:  Merck Millipore EZFITH0LD6
Code : NEW
Additional Details : Weight : 5.80000kg
Product Code. 14872021
  • 8965.00 € / Each
Estimated Shipment: 18-06-2024
to see stock.
Description
Description
  • Vacuum manifolds
  • Support simultaneous filtration of six test samples
  • Made from stainless steel
  • Available with Microfil™ head for use with presterilized, ready-to-use funnels and membranes