missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™-PSQ PVDF Membranes

Product Code. 10491541 Shop All Merck Products
Change view
Click to view available options
Dimensions (LxW):
10 m x 8.5 cm
10 x 10 cm
15 x 15 cm
20 x 20 cm
26 x 26 cm
26.5 cm x 3.75 m
7 x 8.4 cm
8 x 10 cm
8.5 x 13.5 cm
9 x 12 cm
Format:
Roll
Sheet
Unit Size:
1 roll
Each
Pack of 10
Pack of 50
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Format unitSize
10491541 26.5 cm x 3.75 m Roll 1 roll
10153593 7 x 8.4 cm Sheet Pack of 50
11506345 8 x 10 cm Sheet Pack of 10
10163593 8.5 x 13.5 cm Sheet Pack of 10
10146470 10 x 10 cm Sheet Pack of 10
10094450 15 x 15 cm Sheet Pack of 10
10712731 26 x 26 cm Sheet Pack of 10
10481541 20 x 20 cm Sheet Pack of 10
10593984 9 x 12 cm Sheet Pack of 10
16339601 10 m x 8.5 cm Roll Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 10491541 Supplier Merck Supplier No. ISEQ00010
  • ONLINE EXCLUSIVE PRICE
    604.00 €
    554.93 € / 1 rolls
    Save 49.07 €
    8% Off
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For Western blots and protein sequencing

Immobilon-PSQ Membrane:
  • 0.2 μm pore size and a large internal structure
  • Higher protein adsorption and sequencing yields than other membranes
  • Recommended for blotting proteins less than 20 kDa
  • Prevents blow-through of low molecular weight proteins
  • TRUSTED_SUSTAINABILITY

    Specifications

    Length (Metric) 26.5 cm
    Width (Metric) 375 cm
    For Use With (Application) For sequencing low-abundance or low-molecular weight (<20,000 Daltons) proteins
    Width (English) 147.63 in.
    Format Roll
    Material Immobilon-PSQ
    Length (English) 10.43 in.
    Pore Size 0.2 μm
    Dimensions (LxW) 26.5 cm x 3.75 m
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.