missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon™ Western Chemiluminescent HRP Substrate (ECL)

Product Code. 11556345 Shop All Merck Products
Change view
Click to view available options
Quantity:
2 x 25 mL
2 x 50 mL
2 x 250 mL
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11556345 2 x 250 mL Pack of 2
11536345 2 x 25 mL Pack of 2
11546345 2 x 50 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11556345 Supplier Merck Supplier No. WBKLS0500
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Luminol, peroxide solution

Provides high sensitivity over a broad detection range at an economical price

  • Combines high sensitivity and low background
  • Allows detection of high- and low-abundance proteins in the mid-femtogram range
  • Lowers antibody consumption by two- to five-fold compared to less sensitive reagents
  • Compatible with PVDF and nitrocellulose membranes as well as all commonly used buffers and blocking reagents

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Dot/Slot Blot
Quantity 2 x 250 mL
Substrate Horseradish Peroxidase
Sufficient For 5000cm2 Membrane
Content And Storage 250mL luminol, 250mL peroxide solution. 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.