missing translation for 'onlineSavingsMsg'
Learn More

Merck Luminata Forte Western HRP substrate, 100mL

Product Code. 10394675 Shop All Merck Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10394675 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10394675 Supplier Merck Supplier No. WBLUF0100
  • ONLINE EXCLUSIVE PRICE
    220.00 €
    203.11 € / 100mL
    Save 16.89 €
    7% Off
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premixed, ready to use chemiluminescent HRP detection reagent

  • The reagent has been matched to optimize the range and sensitivity of detection using various antibodies in Western blot assays.

  • Premixed for convenience to avoid cross contamination of the reagents.
  • Broad range of sensitivities to satisfy all detection needs.
  • Consistent results with less pipetting error.
  • Stable at 4°C and also at room temperature which allows storage on bench.
  • Best detection performance relative to competition.
  • Compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Applications:

Western blotting, dot/slot blotting

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Storage Requirements 18° to 24°C
For Use With (Application) Dot/Slot Blot, Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.