Learn More
Merck Millicell™ Cell Culture Insert Plates
Product Code. 11700393 Shop All Merck Products
Change view
Click to view available options
Type:
96-well Receiver Tray, Lid
Single-well Feeder Tray, 96-well Receiver Tray, Lid
Single-well Feeder Tray, Lid
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11700393 Single-well Feeder Tray, Lid Pack of 5
11770373 Single-well Feeder Tray, 96-well Receiver Tray, Lid Each
11790383 96-well Receiver Tray, Lid Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11700393 Fornitore Merck N. del fornitore PSHT004S5
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Designed to support cell growth and differentiation of endothelial and epithelial cell lines
  • For easier pipetting and isolateral access
  • Can be used manually or with automated cell seeding, feeding and washing systems
  • Optimized to grow and sustain high-integrity cell monolayers
  • Membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray to reduce the risk of monolayer contamination
  • Tear-drop shaped receiver wells eliminate air bubbles as plates are assembled
  • Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane
Type and number of components vary by model:
PSHT004R1 Includes
  • One 96-Well Cell Culture Plate
  • One Feeder Tray
  • One 96-Well Receiver Tray
  • One Lid
PSHT004R5 Includes
  • Five 96-Well Culture Plates
  • Five Single-Well Feeder Trays
  • Five Lids
PSHT004S5 Includes
  • Five 96-Well Cell Culture Plates
  • Ten 96-Well Receiver Trays
  • Five Lids
  • Cell Attachment
  • Cell Culture
  • Cell Differentiation
  • Drug Transport
  • Permeability
  • Cell-Based Assays
  • Caco-2 Cell Culture
TRUSTED_SUSTAINABILITY
Specifica
Material PCF
Pore Size 0.4 μm
Type Single-well Feeder Tray, Lid
For Use With (Application) Cell Attachment, Cell Culture, Cell Differentiation, Drug Transport
Quantity 5/Pk.
Sterility Sterile
No. of Wells 96
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.