Learn More
Merck Millicell™ Cell Culture Insert Plates
Product Code. 11790383 Shop All Merck Products
Change view
Click to view available options
Type:
96-well Receiver Tray, Lid
Single-well Feeder Tray, 96-well Receiver Tray, Lid
Single-well Feeder Tray, Lid
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11790383 96-well Receiver Tray, Lid Pack of 5
11700393 Single-well Feeder Tray, Lid Pack of 5
11770373 Single-well Feeder Tray, 96-well Receiver Tray, Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11790383 Supplier Merck Supplier No. PSHT004R5
  • ONLINE EXCLUSIVE PRICE
    2520.00 €
    2330.00 € / Pack of 5
    Save 190.00 €
    7% Off
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Designed to support cell growth and differentiation of endothelial and epithelial cell lines
  • For easier pipetting and isolateral access
  • Can be used manually or with automated cell seeding, feeding and washing systems
  • Optimized to grow and sustain high-integrity cell monolayers
  • Membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray to reduce the risk of monolayer contamination
  • Tear-drop shaped receiver wells eliminate air bubbles as plates are assembled
  • Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane
Type and number of components vary by model:
PSHT004R1 Includes
  • One 96-Well Cell Culture Plate
  • One Feeder Tray
  • One 96-Well Receiver Tray
  • One Lid
PSHT004R5 Includes
  • Five 96-Well Culture Plates
  • Five Single-Well Feeder Trays
  • Five Lids
PSHT004S5 Includes
  • Five 96-Well Cell Culture Plates
  • Ten 96-Well Receiver Trays
  • Five Lids
  • Cell Attachment
  • Cell Culture
  • Cell Differentiation
  • Drug Transport
  • Permeability
  • Cell-Based Assays
  • Caco-2 Cell Culture
TRUSTED_SUSTAINABILITY
Specifications
Material PCF
Pore Size 0.4 μm
Type 96-well Receiver Tray, Lid
For Use With (Application) Cell Attachment, Cell Culture, Cell Differentiation, Drug Transport
Quantity 5/Pk.
Sterility Sterile
No. of Wells 96