missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ 96-Well Assay Plates for Receptor Ligand Binding Assays

Ideal for performing receptor binding assays involving whole cells, membrane fragments, or purified receptors; also suitable for Scatchard analysis, as has been demonstrated in numerous receptor-binding assays including Interleukin I, Angiotensin II, and C5a studies

Brand:  Merck MABVN0B50

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 10701971

  • 2550.00 € / Pack of 50
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

  • Nontoxic
  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold

Choice of filter materials

  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes

Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.

Specifications

Specifications

Standard
8 x 12
Opaque
Barex
96
0.28 cm2
50/Pk.
1.28 cm
1.44 cm
Hydrophilic PVDF
1.2 μm
MultiScreen-BV Plate
0.05 to 0.25 mL
Non-sterile
Receptor binding assays, resin, bead based assays
0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44 cm)
8.55 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers