Learn More
Merck MultiScreenHTS 384-Well Filter Plates
Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.
Brand: Merck MZHVN0W10
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.- Rigid sidewalls allows full compatibility with automated/robotic systems
- Unique plate design and individually sealed filter discs prevent cross-talk between wells
- Opaque plastic lids prevents light transfer between wells during direct scintillation counting
- Barcoding available
- Meet ANSI™/SBS standards
Specifications
Standard | |
16 x 24 | |
Opaque, White Bottom | |
384 | |
0.09cm2 | |
10/Pk. | |
1.28 cm | |
1.44 cm |
Durapore PVDF | |
0.45 μm | |
Styrene | |
Non-sterile | |
Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays | |
1.28 x 8.55 x 1.44 cm | |
8.55 cm |