missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS 384-Well Filter Plates
Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.
1185.00 € - 5325.00 €
Specifications
Format | Standard |
---|---|
Membrane | Glass Fiber, Polyester Mesh |
Array | 16 x 24 |
Color | Opaque, White Bottom |
Material | Barex |
Product Code | Brand | Membrane | Pore Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Membrane | Pore Size | Quantity | Price | Quantity & Availability | |||
10500275
|
Merck
MZFCN0W10 |
Glass Fiber, Polyester Mesh | 1.2 μm | 10/Pk. |
1185.00 €
Pack of 10 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||
11542144
|
Merck
MZFBN0W50 |
Glass Fiber, Polyester Mesh | 1 μm | 50/Pk. |
5325.00 €
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||
10602605
|
Merck
MZFBN0W10 |
Glass Fiber, Polyester Mesh | 1 μm | 10/Pk. |
1185.00 €
Pack of 10 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||
11784694
|
Merck
MZFCN0W50 |
Glass Fiber, Polyester Mesh | 1.2 μm | 50/Pk. |
5325.00 €
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||
Description
For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.- Rigid sidewalls allows full compatibility with automated/robotic systems
- Unique plate design and individually sealed filter discs prevent cross-talk between wells
- Opaque plastic lids prevents light transfer between wells during direct scintillation counting
- Barcoding available
- Meet ANSI™/SBS standards
Specifications
Standard | |
16 x 24 | |
Barex | |
Non-sterile | |
1.28 cm | |
1.44 cm |
Glass Fiber, Polyester Mesh | |
Opaque, White Bottom | |
384 | |
1.28 x 8.55 x 1.44 cm | |
8.55 cm |