missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS 96-Well Filter Plates
Receptor/ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification
Brand: Merck MSFBNXB50
Code : NEW
Additional Details : Weight : 0.10000kg
Specifications
ANSI, SBS | |
Polyester Mesh | |
1 μm | |
Opaque | |
96 | |
0.26 cm2 | |
0.5 x 8.55 x 0.56 in. (1.28 x 21.71 x 1.44 cm) | |
21.71 cm |
Standard | |
8 x 12 | |
Low Binding | |
Barex | |
Non-sterile | |
Receptor, ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification | |
1.28 cm | |
1.44 cm |