missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Pipe Thread Fittings
Alle ansehen Merck Produkte
Click to view available options
Type:
Pipe Thread Fitting
Pipe Thread Valve
Connection Type:
1/4 in. NPTM
1/4 in. NPTM Close Nipple
1/8 in. -1/8 in. NPTM 3-way Plug Valve
Packungsgröße:
Each
Beschreibung
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Spezifikation
Spezifikation
| Connection Type | 1/4 in. NPTM |
| For Use With (Equipment) | Water Purification System |
| Type | Pipe Thread Fitting |
| Material | Series 300 stainless steel |
| Diameter (English) Inside | 0.25 in. |
| Diameter (Metric) Inside | 0.63 cm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur